Assay Details
| Gene Symbol: Washc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252475.1 | XP_021108134.1 | 12 - 13 | 1439 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Washc2c| Gene Symbol: Washc2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252475.1 | XP_021108134.1 | 12 - 13 | 1439 | 93 | |
Set Membership: |
|