Assay Details
Gene Symbol: LOC106008944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021254099.1 | XP_021109758.1 | 2 - 3 | 450 | 91 | |
XM_021254100.1 | XP_021109759.1 | 2 - 3 | 290 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
