Assay Details
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004836481.3 | XP_004836538.1 | 5 - 6 | 785 | 80 | |
XM_021240520.1 | XP_021096179.1 | 6 - 7 | 763 | 80 | ||
XM_021240526.1 | XP_021096185.1 | 5 - 6 | 743 | 80 | ||
XM_021240530.1 | XP_021096189.1 | 4 - 5 | 526 | 80 | ||
XM_021240538.1 | XP_021096197.1 | 5 - 6 | 634 | 80 | ||
XM_021240544.1 | XP_021096203.1 | 6 - 7 | 669 | 80 | ||
XM_021240553.1 | XP_021096212.1 | 5 - 6 | 655 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
