Assay Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839659.3 | XP_004839716.1 | 5 - 6 | 574 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
