Assay Details
Gene Symbol: Stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840666.2 | XP_004840723.1 | 2 - 3 | 1103 | 77 | |
XM_013070685.2 | XP_012926139.1 | 2 - 3 | 657 | 77 | ||
XM_021254970.1 | XP_021110629.1 | 2 - 3 | 1103 | 77 | ||
XM_021254971.1 | XP_021110630.1 | 2 - 3 | 671 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
