Assay Details
Gene Symbol: Bean1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843110.3 | XP_004843167.1 | 2 - 3 | 198 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004843110.3 transcripts.
Set Membership: |
![]() ![]() |
