Assay Details
Gene Symbol: CUNH4orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847398.3 | XP_004847455.1 | 3 - 4 | 326 | 85 | |
XM_013073145.2 | XP_012928599.1 | 3 - 4 | 321 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
