Assay Details
| Gene Symbol: C2cd4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854108.2 | XP_004854165.1 | 1 | 824 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2cd4d| Gene Symbol: C2cd4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854108.2 | XP_004854165.1 | 1 | 824 | 54 | |
Set Membership: |
|