Assay Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858688.3 | XP_004858745.1 | 3 - 4 | 583 | 68 | |
| XM_004858689.3 | XP_004858746.1 | 3 - 4 | 552 | 68 | ||
| XM_004858690.3 | XP_004858747.1 | 3 - 4 | 515 | 68 | ||
| XM_004858692.3 | XP_004858749.1 | 3 - 4 | 599 | 68 | ||
| XM_021239613.1 | XP_021095272.1 | 3 - 4 | 457 | 68 | ||
More Information
Set Membership: |
|

Nit1
Genomic Map