Assay Details
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859905.2 | XP_004859962.1 | 8 - 9 | 904 | 79 | |
XM_021240513.1 | XP_021096172.1 | 8 - 9 | 904 | 79 | ||
XM_021240514.1 | XP_021096173.1 | 6 - 7 | 682 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
