Assay Details
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869518.3 | XP_004869575.1 | 7 - 8 | 1094 | 63 | |
XM_021248085.1 | XP_021103744.1 | 8 - 9 | 1238 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
