Assay Details
Gene Symbol: LOC101709173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872177.2 | XP_004872234.1 | 10 - 11 | 2733 | 107 | |
XM_004872178.3 | XP_004872235.1 | 10 - 11 | 2548 | 107 | ||
XM_013068293.2 | XP_012923747.1 | 8 - 9 | 2560 | 107 | ||
XM_013068294.2 | XP_012923748.1 | 8 - 9 | 2383 | 107 | ||
XM_021250166.1 | XP_021105825.1 | 10 - 11 | 2560 | 107 | ||
XM_021250167.1 | XP_021105826.1 | 9 - 10 | 2503 | 107 | ||
XM_021250168.1 | XP_021105827.1 | 9 - 10 | 2430 | 107 | ||
XM_021250170.1 | XP_021105829.1 | 9 - 10 | 2615 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
