Assay Details
Gene Symbol: LOC101709173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872177.2 | XP_004872234.1 | 5 - 6 | 813 | 81 | |
XM_004872178.3 | XP_004872235.1 | 5 - 6 | 628 | 81 | ||
XM_013068293.2 | XP_012923747.1 | 3 - 4 | 640 | 81 | ||
XM_013068294.2 | XP_012923748.1 | 3 - 4 | 463 | 81 | ||
XM_021250166.1 | XP_021105825.1 | 5 - 6 | 640 | 81 | ||
XM_021250167.1 | XP_021105826.1 | 4 - 5 | 583 | 81 | ||
XM_021250168.1 | XP_021105827.1 | 4 - 5 | 510 | 81 | ||
XM_021250170.1 | XP_021105829.1 | 4 - 5 | 695 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
