Assay Details
Gene Symbol: LOC101709173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872177.2 | XP_004872234.1 | 7 - 8 | 2208 | 105 | |
XM_004872178.3 | XP_004872235.1 | 7 - 8 | 2023 | 105 | ||
XM_013068293.2 | XP_012923747.1 | 5 - 6 | 2035 | 105 | ||
XM_013068294.2 | XP_012923748.1 | 5 - 6 | 1858 | 105 | ||
XM_021250166.1 | XP_021105825.1 | 7 - 8 | 2035 | 105 | ||
XM_021250167.1 | XP_021105826.1 | 6 - 7 | 1978 | 105 | ||
XM_021250168.1 | XP_021105827.1 | 6 - 7 | 1905 | 105 | ||
XM_021250170.1 | XP_021105829.1 | 6 - 7 | 2090 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
