Assay Details
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013073342.2 | XP_012928796.1 | 3 - 4 | 1033 | 72 | |
XM_013073343.2 | XP_012928797.1 | 4 - 5 | 1051 | 72 | ||
XM_013073344.2 | XP_012928798.1 | 3 - 4 | 1030 | 72 | ||
XM_013073345.2 | XP_012928799.1 | 3 - 4 | 965 | 72 | ||
XM_021260921.1 | XP_021116580.1 | 3 - 4 | 1423 | 72 | ||
XM_021260922.1 | XP_021116581.1 | 4 - 5 | 1065 | 72 | ||
XM_021260923.1 | XP_021116582.1 | 3 - 4 | 1423 | 72 | ||
XM_021260924.1 | XP_021116583.1 | 3 - 4 | 1423 | 72 | ||
XM_021260925.1 | XP_021116584.1 | 3 - 4 | 1423 | 72 | ||
XM_021260926.1 | XP_021116585.1 | 3 - 4 | 1423 | 72 | ||
XM_021260927.1 | XP_021116586.1 | 2 - 3 | 388 | 72 | ||
XM_021260928.1 | XP_021116587.1 | 3 - 4 | 412 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
