Assay Details
Gene Symbol: LOC101704976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077541.2 | XP_012932995.1 | 1 - 2 | 479 | 105 | |
XM_013077542.2 | XP_012932996.1 | 1 - 2 | 479 | 105 | ||
XM_021239673.1 | XP_021095332.1 | 2 - 3 | 364 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013077541.2, XM_013077542.2, XM_021239673.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
