Assay Details
Gene Symbol: LOC101704976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077541.2 | XP_012932995.1 | 5 - 6 | 1389 | 65 | |
XM_021239672.1 | XP_021095331.1 | 6 - 7 | 1616 | 65 | ||
XM_021239673.1 | XP_021095332.1 | 6 - 7 | 1346 | 65 | ||
XM_021239674.1 | XP_021095333.1 | 7 - 8 | 1514 | 65 | ||
XM_021239675.1 | XP_021095334.1 | 6 - 7 | 1333 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
