Assay Details
Gene Symbol: LOC101704976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077541.2 | XP_012932995.1 | 6 - 7 | 1808 | 120 | |
XM_013077542.2 | XP_012932996.1 | 5 - 6 | 1487 | 120 | ||
XM_021239672.1 | XP_021095331.1 | 7 - 8 | 2035 | 120 | ||
XM_021239673.1 | XP_021095332.1 | 7 - 8 | 1765 | 120 | ||
XM_021239674.1 | XP_021095333.1 | 8 - 9 | 1933 | 120 | ||
XM_021239675.1 | XP_021095334.1 | 7 - 8 | 1752 | 120 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
