Assay Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021266443.1 | XP_021122102.1 | 1 - 2 | 390 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il2ra| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021266443.1 | XP_021122102.1 | 1 - 2 | 390 | 69 | |
Set Membership: |
|