Assay Details
| Gene Symbol: LOC101715399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838650.2 | XP_004838707.1 | 4 - 5 | 726 | 101 | |
| XM_021253187.1 | XP_021108846.1 | 4 - 5 | 551 | 101 | ||
More Information
Set Membership: |
|

LOC101715399
Genomic Map