Assay Details
Gene Symbol: LOC101715399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838650.2 | XP_004838707.1 | 7 - 8 | 1131 | 112 | |
XM_021253187.1 | XP_021108846.1 | 7 - 8 | 956 | 112 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
