Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 10 - 11 | 1405 | 88 | |
XM_004839795.3 | XP_004839852.2 | 9 - 10 | 1214 | 88 | ||
XM_004839796.3 | XP_004839853.2 | 8 - 9 | 950 | 88 | ||
XM_013070363.2 | XP_012925817.1 | 10 - 11 | 1407 | 88 | ||
XM_013070364.2 | XP_012925818.1 | 7 - 8 | 762 | 88 | ||
XM_013070365.2 | XP_012925819.1 | 6 - 7 | 839 | 88 | ||
XM_013070366.2 | XP_012925820.1 | 10 - 11 | 1414 | 88 | ||
XM_021254020.1 | XP_021109679.1 | 4 - 5 | 449 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
