Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 11 - 12 | 1540 | 76 | |
XM_004839795.3 | XP_004839852.2 | 10 - 11 | 1349 | 76 | ||
XM_004839796.3 | XP_004839853.2 | 9 - 10 | 1085 | 76 | ||
XM_013070363.2 | XP_012925817.1 | 11 - 12 | 1542 | 76 | ||
XM_013070364.2 | XP_012925818.1 | 8 - 9 | 897 | 76 | ||
XM_013070365.2 | XP_012925819.1 | 7 - 8 | 974 | 76 | ||
XM_013070366.2 | XP_012925820.1 | 11 - 12 | 1549 | 76 | ||
XM_021254020.1 | XP_021109679.1 | 5 - 6 | 584 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
