Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 15 - 16 | 2247 | 144 | |
XM_004839795.3 | XP_004839852.2 | 14 - 15 | 2056 | 144 | ||
XM_004839796.3 | XP_004839853.2 | 13 - 14 | 1792 | 144 | ||
XM_013070363.2 | XP_012925817.1 | 15 - 16 | 2249 | 144 | ||
XM_013070364.2 | XP_012925818.1 | 12 - 13 | 1604 | 144 | ||
XM_013070365.2 | XP_012925819.1 | 11 - 12 | 1681 | 144 | ||
XM_013070366.2 | XP_012925820.1 | 15 - 16 | 2256 | 144 | ||
XM_013070367.2 | XP_012925821.1 | 5 - 6 | 764 | 144 | ||
XM_021254020.1 | XP_021109679.1 | 9 - 10 | 1291 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
