Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 16 - 17 | 2390 | 73 | |
XM_004839795.3 | XP_004839852.2 | 15 - 16 | 2199 | 73 | ||
XM_004839796.3 | XP_004839853.2 | 14 - 15 | 1935 | 73 | ||
XM_013070363.2 | XP_012925817.1 | 16 - 17 | 2392 | 73 | ||
XM_013070364.2 | XP_012925818.1 | 13 - 14 | 1747 | 73 | ||
XM_013070365.2 | XP_012925819.1 | 12 - 13 | 1824 | 73 | ||
XM_013070366.2 | XP_012925820.1 | 16 - 17 | 2399 | 73 | ||
XM_013070367.2 | XP_012925821.1 | 6 - 7 | 907 | 73 | ||
XM_021254020.1 | XP_021109679.1 | 10 - 11 | 1434 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
