Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 19 - 20 | 2651 | 90 | |
XM_004839795.3 | XP_004839852.2 | 19 - 20 | 2541 | 90 | ||
XM_004839796.3 | XP_004839853.2 | 18 - 19 | 2277 | 90 | ||
XM_013070363.2 | XP_012925817.1 | 20 - 21 | 2734 | 90 | ||
XM_013070364.2 | XP_012925818.1 | 17 - 18 | 2089 | 90 | ||
XM_013070365.2 | XP_012925819.1 | 16 - 17 | 2166 | 90 | ||
XM_013070366.2 | XP_012925820.1 | 20 - 21 | 2741 | 90 | ||
XM_013070367.2 | XP_012925821.1 | 10 - 11 | 1249 | 90 | ||
XM_013070369.2 | XP_012925823.1 | 2 - 3 | 270 | 90 | ||
XM_021254020.1 | XP_021109679.1 | 14 - 15 | 1776 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013070369.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
