Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 20 - 21 | 2864 | 104 | |
XM_004839795.3 | XP_004839852.2 | 20 - 21 | 2754 | 104 | ||
XM_004839796.3 | XP_004839853.2 | 19 - 20 | 2490 | 104 | ||
XM_013070363.2 | XP_012925817.1 | 21 - 22 | 2947 | 104 | ||
XM_013070364.2 | XP_012925818.1 | 18 - 19 | 2302 | 104 | ||
XM_013070365.2 | XP_012925819.1 | 17 - 18 | 2379 | 104 | ||
XM_013070366.2 | XP_012925820.1 | 21 - 22 | 2954 | 104 | ||
XM_013070367.2 | XP_012925821.1 | 11 - 12 | 1462 | 104 | ||
XM_013070369.2 | XP_012925823.1 | 3 - 4 | 483 | 104 | ||
XM_021254020.1 | XP_021109679.1 | 15 - 16 | 1989 | 104 | ||
XM_021254021.1 | XP_021109680.1 | 2 - 3 | 354 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
