Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 26 - 27 | 3812 | 104 | |
XM_004839795.3 | XP_004839852.2 | 26 - 27 | 3702 | 104 | ||
XM_004839796.3 | XP_004839853.2 | 25 - 26 | 3438 | 104 | ||
XM_013070363.2 | XP_012925817.1 | 27 - 28 | 3895 | 104 | ||
XM_013070364.2 | XP_012925818.1 | 24 - 25 | 3250 | 104 | ||
XM_013070365.2 | XP_012925819.1 | 23 - 24 | 3327 | 104 | ||
XM_013070367.2 | XP_012925821.1 | 17 - 18 | 2410 | 104 | ||
XM_013070369.2 | XP_012925823.1 | 9 - 10 | 1431 | 104 | ||
XM_021254020.1 | XP_021109679.1 | 21 - 22 | 2937 | 104 | ||
XM_021254021.1 | XP_021109680.1 | 8 - 9 | 1302 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
