Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 5 - 6 | 711 | 87 | |
XM_004839796.3 | XP_004839853.2 | 3 - 4 | 256 | 87 | ||
XM_013070363.2 | XP_012925817.1 | 5 - 6 | 713 | 87 | ||
XM_013070364.2 | XP_012925818.1 | 2 - 3 | 68 | 87 | ||
XM_013070366.2 | XP_012925820.1 | 5 - 6 | 720 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013070364.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
