Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 6 - 7 | 885 | 67 | |
XM_004839795.3 | XP_004839852.2 | 5 - 6 | 694 | 67 | ||
XM_004839796.3 | XP_004839853.2 | 4 - 5 | 430 | 67 | ||
XM_013070363.2 | XP_012925817.1 | 6 - 7 | 887 | 67 | ||
XM_013070364.2 | XP_012925818.1 | 3 - 4 | 242 | 67 | ||
XM_013070365.2 | XP_012925819.1 | 2 - 3 | 319 | 67 | ||
XM_013070366.2 | XP_012925820.1 | 6 - 7 | 894 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
