Assay Details
Gene Symbol: LOC101717251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004839793.3 | XP_004839850.1 | 7 - 8 | 1009 | 104 | |
XM_004839795.3 | XP_004839852.2 | 6 - 7 | 818 | 104 | ||
XM_004839796.3 | XP_004839853.2 | 5 - 6 | 554 | 104 | ||
XM_013070363.2 | XP_012925817.1 | 7 - 8 | 1011 | 104 | ||
XM_013070364.2 | XP_012925818.1 | 4 - 5 | 366 | 104 | ||
XM_013070365.2 | XP_012925819.1 | 3 - 4 | 443 | 104 | ||
XM_013070366.2 | XP_012925820.1 | 7 - 8 | 1018 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
