Assay Details
Gene Symbol: Kiaa0895l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843056.3 | XP_004843113.1 | 5 - 6 | 1521 | 56 | |
XM_004843057.2 | XP_004843114.1 | 5 - 6 | 1556 | 56 | ||
XM_021256898.1 | XP_021112557.1 | 5 - 6 | 1819 | 56 | ||
XM_021256899.1 | XP_021112558.1 | 5 - 6 | 1820 | 56 | ||
XM_021256901.1 | XP_021112560.1 | 6 - 7 | 1343 | 56 | ||
XM_021256902.1 | XP_021112561.1 | 6 - 7 | 1085 | 56 | ||
XM_021256903.1 | XP_021112562.1 | 6 - 7 | 1050 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
