Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846806.3 | XP_004846863.1 | 3 - 4 | 1190 | 64 | |
XM_021260026.1 | XP_021115685.1 | 3 - 4 | 1075 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
