Assay Details
Gene Symbol: LOC101721703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848051.3 | XP_004848108.1 | 13 - 14 | 2208 | 106 | |
XM_021261053.1 | XP_021116712.1 | 12 - 13 | 1631 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
