Assay Details
Gene Symbol: LOC101721703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848051.3 | XP_004848108.1 | 2 - 3 | 777 | 82 | |
XM_021261053.1 | XP_021116712.1 | 1 - 2 | 200 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
