Assay Details
Gene Symbol: Opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853473.3 | XP_004853530.1 | 3 - 4 | 1003 | 89 | |
XM_021265485.1 | XP_021121144.1 | 4 - 5 | 1045 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
