Assay Details
Gene Symbol: Bicd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855021.3 | XP_004855078.1 | 2 - 3 | 554 | 56 | |
XM_004855022.3 | XP_004855079.1 | 2 - 3 | 554 | 56 | ||
XM_004855023.3 | XP_004855080.1 | 2 - 3 | 554 | 56 | ||
XM_004855024.3 | XP_004855081.1 | 2 - 3 | 554 | 56 | ||
XM_004855025.3 | XP_004855082.1 | 2 - 3 | 554 | 56 | ||
XM_004855026.3 | XP_004855083.1 | 2 - 3 | 554 | 56 | ||
XM_004855027.3 | XP_004855084.1 | 2 - 3 | 554 | 56 | ||
XM_013075976.2 | XP_012931430.1 | 2 - 3 | 554 | 56 | ||
XM_013075977.2 | XP_012931431.1 | 2 - 3 | 554 | 56 | ||
XM_021236545.1 | XP_021092204.1 | 2 - 3 | 554 | 56 | ||
XM_021236547.1 | XP_021092206.1 | 2 - 3 | 554 | 56 | ||
XM_021236548.1 | XP_021092207.1 | 2 - 3 | 554 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
