Assay Details
| Gene Symbol: Ctif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855117.2 | XP_004855174.1 | 5 - 6 | 658 | 61 | |
| XM_004855118.2 | XP_004855175.1 | 5 - 6 | 697 | 61 | ||
| XM_004855119.3 | XP_004855176.1 | 5 - 6 | 689 | 61 | ||
| XM_004855120.3 | XP_004855177.1 | 5 - 6 | 499 | 61 | ||
| XM_004855121.2 | XP_004855178.1 | 5 - 6 | 658 | 61 | ||
| XM_004855122.2 | XP_004855179.1 | 5 - 6 | 658 | 61 | ||
More Information
Set Membership: |
|

Ctif
Genomic Map