Assay Details
Gene Symbol: Traf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857013.3 | XP_004857070.2 | 6 - 7 | 1380 | 66 | |
XM_004857014.2 | XP_004857071.1 | 6 - 7 | 888 | 66 | ||
XM_013076892.2 | XP_012932346.1 | 6 - 7 | 909 | 66 | ||
XM_021238240.1 | XP_021093899.1 | 6 - 7 | 1358 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
