Assay Details
Gene Symbol: Ddit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861228.3 | XP_004861285.1 | 3 - 4 | 368 | 92 | |
XM_004861229.3 | XP_004861286.1 | 3 - 4 | 526 | 92 | ||
XM_013078272.2 | XP_012933726.1 | 3 - 4 | 464 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
