Assay Details
| Gene Symbol: Kiss1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865994.2 | XP_004866051.1 | 4 - 5 | 890 | 84 | |
| XM_021245156.1 | XP_021100815.1 | 4 - 5 | 807 | 84 | ||
More Information
Set Membership: |
|

Kiss1r
Genomic Map