Assay Details
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870351.3 | XP_004870408.1 | 8 - 9 | 1391 | 76 | |
XM_021248702.1 | XP_021104361.1 | 8 - 9 | 1398 | 76 | ||
XM_021248703.1 | XP_021104362.1 | 7 - 8 | 1310 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
