Assay Details
| Gene Symbol: Nthl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873658.2 | XP_004873715.1 | 1 - 2 | 135 | 69 | |
| XM_004873659.2 | XP_004873716.1 | 1 - 2 | 134 | 69 | ||
| XM_004873660.2 | XP_004873717.1 | 1 - 2 | 136 | 69 | ||
| XM_004873662.2 | XP_004873719.1 | 1 - 2 | 135 | 69 | ||
| XM_021251312.1 | XP_021106971.1 | 1 - 2 | 136 | 69 | ||
| XM_021251313.1 | XP_021106972.1 | 1 - 2 | 135 | 69 | ||
| XM_021251314.1 | XP_021106973.1 | 1 - 2 | 137 | 69 | ||
| XM_021251316.1 | XP_021106975.1 | 1 - 2 | 137 | 69 | ||
More Information
Set Membership: |
|

Nthl1
Genomic Map