Assay Details
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068127.2 | XP_012923581.1 | 4 - 5 | 279 | 63 | |
XM_021249825.1 | XP_021105484.1 | 3 - 4 | 472 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
