Assay Details
Gene Symbol: Sapcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021261626.1 | XP_021117285.1 | 3 - 4 | 1587 | 85 | |
XM_021261627.1 | XP_021117286.1 | 3 - 4 | 1582 | 85 | ||
XM_021261628.1 | XP_021117287.1 | 2 - 3 | 628 | 85 | ||
XM_021261629.1 | XP_021117288.1 | 2 - 3 | 628 | 85 | ||
XM_021261630.1 | XP_021117289.1 | 3 - 4 | 1596 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
