Assay Details
Gene Symbol: LOC101696935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840505.2 | XP_004840562.1 | 2 - 3 | 628 | 90 | |
XM_004840506.3 | XP_004840563.1 | 2 - 3 | 747 | 90 | ||
XM_004840507.2 | XP_004840564.1 | 2 - 3 | 628 | 90 | ||
XM_021254806.1 | XP_021110465.1 | 2 - 3 | 649 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
