Assay Details
Gene Symbol: LOC101696935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840505.2 | XP_004840562.1 | 6 - 7 | 1799 | 81 | |
XM_004840506.3 | XP_004840563.1 | 6 - 7 | 1918 | 81 | ||
XM_004840507.2 | XP_004840564.1 | 5 - 6 | 1697 | 81 | ||
XM_021254806.1 | XP_021110465.1 | 6 - 7 | 1820 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
