Assay Details
| Gene Symbol: LOC101696935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840505.2 | XP_004840562.1 | 8 - 9 | 2098 | 78 | |
| XM_004840506.3 | XP_004840563.1 | 8 - 9 | 2217 | 78 | ||
| XM_004840507.2 | XP_004840564.1 | 7 - 8 | 1996 | 78 | ||
| XM_021254806.1 | XP_021110465.1 | 8 - 9 | 2119 | 78 | ||
More Information
Set Membership: |
|

LOC101696935
Genomic Map