Assay Details
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847688.3 | XP_004847745.1 | 4 - 5 | 906 | 103 | |
XM_004847689.3 | XP_004847746.1 | 4 - 5 | 886 | 103 | ||
XM_004847690.3 | XP_004847747.1 | 4 - 5 | 887 | 103 | ||
XM_004847691.3 | XP_004847748.1 | 3 - 4 | 803 | 103 | ||
XM_021260649.1 | XP_021116308.1 | 5 - 6 | 843 | 103 | ||
XM_021260650.1 | XP_021116309.1 | 3 - 4 | 824 | 103 | ||
XM_021260651.1 | XP_021116310.1 | 3 - 4 | 392 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
