Assay Details
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862163.2 | XP_004862220.1 | 8 - 9 | 1279 | 58 | |
XM_004862164.3 | XP_004862221.1 | 7 - 8 | 1257 | 58 | ||
XM_021242434.1 | XP_021098093.1 | 6 - 7 | 1150 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
